Assay Details
| Gene Symbol: CID7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336050.1 | NP_001324896.1 | 6 - 7 | 1808 | 91 | |
| NM_128185.3 | NP_180196.1 | 6 - 7 | 1667 | 91 | ||
| GenBank mRNA | BT030338.1 | - | 4 - 5 | 1409 | 91 | |
More Information
Set Membership: |
|

CID7
Genomic Map