Assay Details
| Gene Symbol: HEMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336073.1 | NP_001324266.1 | 1 - 2 | 388 | 89 | |
| NM_001336074.1 | NP_001324267.1 | 1 - 2 | 389 | 89 | ||
| NM_128211.4 | NP_565625.1 | 1 - 2 | 388 | 89 | ||
| GenBank mRNA | BT026116.1 | - | 1 - 2 | 252 | 89 | |
More Information
Set Membership: |
|

HEMD
Genomic Map