Assay Details
| Gene Symbol: BAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128228.4 | NP_180239.1 | 3 - 4 | 1358 | 97 | |
| GenBank mRNA | AK175232.1 | - | 3 - 4 | 1195 | 97 | |
| AK229743.1 | - | 3 - 4 | 1195 | 97 | ||
| BT010564.1 | - | 3 - 4 | 1121 | 97 | ||
More Information
Set Membership: |
|

BAS1
Genomic Map