Assay Details
| Gene Symbol: CYCB1;4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128233.3 | NP_180244.1 | 3 - 4 | 743 | 71 | |
| GenBank mRNA | AY050781.1 | - | 3 - 4 | 532 | 71 | |
| BT015043.1 | - | 3 - 4 | 310 | 71 | ||
More Information
Set Membership: |
|

CYCB1;4
Genomic Map