Assay Details
| Gene Symbol: emb1187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128240.4 | NP_180251.1 | 1 - 2 | 482 | 76 | |
| GenBank mRNA | AK229423.1 | - | 1 - 2 | 132 | 76 | |
| AX765810.1 | - | 1 - 2 | 124 | 76 | ||
More Information
Set Membership: |
|

emb1187
Genomic Map