Assay Details
| Gene Symbol: SE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128268.3 | NP_565635.1 | 3 - 4 | 760 | 111 | |
| GenBank mRNA | AF311221.1 | - | 3 - 4 | 594 | 111 | |
| AF428305.1 | - | 3 - 4 | 599 | 111 | ||
| AY039915.1 | - | 3 - 4 | 606 | 111 | ||
| AY142569.1 | - | 3 - 4 | 570 | 111 | ||
| AY143937.1 | - | 3 - 4 | 570 | 111 | ||
More Information
Set Membership: |
|

SE
Genomic Map