Assay Details
Gene Symbol: PAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128277.5 | NP_180287.2 | 6 - 7 | 1392 | 108 | |
GenBank mRNA | AY065067.1 | - | 6 - 7 | 1228 | 108 | |
AY133599.1 | - | 6 - 7 | 1212 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
