Assay Details
Gene Symbol: AT2G27430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128303.4 | NP_180312.2 | 1 - 2 | 659 | 76 | |
GenBank mRNA | AY169003.1 | - | 1 - 2 | 498 | 76 | |
AY954802.1 | - | 1 - 2 | 458 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
