Assay Details
| Gene Symbol: NF-YB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128307.6 | NP_180316.1 | 1 - 2 | 252 | 86 | |
| GenBank mRNA | BT004083.1 | - | 1 - 2 | 182 | 86 | |
More Information
Set Membership: |
|

NF-YB11
Genomic Map