Assay Details
Gene Symbol: SKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128319.4 | NP_180328.1 | 2 - 3 | 919 | 63 | |
GenBank mRNA | AF367297.1 | - | 2 - 3 | 767 | 63 | |
AY091684.1 | - | 2 - 3 | 589 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
