Assay Details
| Gene Symbol: AT2G27980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128357.7 | NP_180365.6 | 4 - 5 | 1871 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G27980| Gene Symbol: AT2G27980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128357.7 | NP_180365.6 | 4 - 5 | 1871 | 107 | |
Set Membership: |
|