Assay Details
| Gene Symbol: CSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128379.4 | NP_565666.1 | 1 - 2 | 401 | 101 | |
| GenBank mRNA | AK227096.1 | - | 1 - 2 | 312 | 101 | |
| AY064050.1 | - | 1 - 2 | 311 | 101 | ||
| AY133756.1 | - | 1 - 2 | 265 | 101 | ||
More Information
Set Membership: |
|

CSD2
Genomic Map