Assay Details
| Gene Symbol: DRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128398.4 | NP_565672.1 | 2 - 3 | 798 | 61 | |
| GenBank mRNA | AY034976.1 | - | 2 - 3 | 723 | 61 | |
| AY062965.1 | - | 2 - 3 | 472 | 61 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length less than or equal to 70  3' Most  Detect Genomic DNA  Recommended Assay  Arabidopsis thaliana  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 DRB2
DRB2 Genomic Map
Genomic Map Back To Top
 Back To Top