Assay Details
Gene Symbol: PRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128422.3 | NP_180429.1 | 1 - 2 | 545 | 71 | |
GenBank mRNA | AB058920.1 | - | 1 - 2 | 545 | 71 | |
AY251397.1 | - | 1 - 2 | 443 | 71 | ||
DQ446575.1 | - | 1 - 2 | 443 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
