Assay Details
Gene Symbol: PLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128448.4 | NP_180455.1 | 1 - 2 | 1654 | 102 | |
GenBank mRNA | AY065299.1 | - | 1 - 2 | 1653 | 102 | |
AY096450.1 | - | 1 - 2 | 1477 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
