Assay Details
| Gene Symbol: AT3G21360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113031.2 | NP_188773.1 | 2 - 3 | 688 | 92 | |
| GenBank mRNA | AK117772.1 | - | 2 - 3 | 688 | 92 | |
| BT031354.1 | - | 2 - 3 | 627 | 92 | ||
More Information
Set Membership: |
|

AT3G21360
Genomic Map