Assay Details
Gene Symbol: CERK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113058.4 | NP_566689.2 | 4 - 5 | 974 | 106 | |
GenBank mRNA | AB367524.1 | - | 4 - 5 | 833 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
