Assay Details
Gene Symbol: HYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113071.2 | NP_188813.1 | 1 | 1542 | 142 | |
GenBank mRNA | AF372973.1 | - | 1 | 1493 | 142 | |
AF428321.1 | - | 1 | 1542 | 142 | ||
AY140044.1 | - | 1 | 1518 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
