Assay Details
| Gene Symbol: ATXR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113077.5 | NP_188819.2 | 11 - 12 | 1289 | 74 | |
| GenBank mRNA | AK221711.1 | - | 11 - 12 | 1226 | 74 | |
| BT020260.1 | - | 11 - 12 | 1173 | 74 | ||
| BT021112.1 | - | 11 - 12 | 1166 | 74 | ||
More Information
Set Membership: |
|

ATXR2
Genomic Map