Assay Details
| Gene Symbol: BBX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338552.1 | NP_001319614.1 | 1 - 2 | 487 | 138 | |
| NM_001338553.1 | NP_001326646.1 | 2 - 3 | 764 | 138 | ||
| NM_113084.2 | NP_188826.2 | 1 - 2 | 552 | 138 | ||
| GenBank mRNA | AB493626.1 | - | 1 - 2 | 487 | 138 | |
More Information
Set Membership: |
|

BBX10
Genomic Map