Assay Details
| Gene Symbol: POP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203018.1 | NP_001189947.1 | 14 - 15 | 1407 | 119 | |
| NM_113117.4 | NP_566700.1 | 14 - 15 | 1204 | 119 | ||
| GenBank mRNA | AY034923.1 | - | 14 - 15 | 1195 | 119 | |
| AY142571.1 | - | 14 - 15 | 1067 | 119 | ||
More Information
Set Membership: |
|

POP2
Genomic Map