Assay Details
Gene Symbol: POP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203018.1 | NP_001189947.1 | 18 - 19 | 1771 | 69 | |
NM_113117.4 | NP_566700.1 | 18 - 19 | 1568 | 69 | ||
GenBank mRNA | AY034923.1 | - | 18 - 19 | 1559 | 69 | |
AY142571.1 | - | 18 - 19 | 1431 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
