Assay Details
| Gene Symbol: AT3G22620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113160.3 | NP_566713.1 | 2 - 3 | 439 | 86 | |
| GenBank mRNA | AY035005.1 | - | 2 - 3 | 439 | 86 | |
| AY113897.1 | - | 2 - 3 | 358 | 86 | ||
More Information
Set Membership: |
|

AT3G22620
Genomic Map