Assay Details
Gene Symbol: LFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113199.5 | NP_566721.1 | 7 - 8 | 1204 | 87 | |
GenBank mRNA | AY072191.1 | - | 7 - 8 | 1155 | 87 | |
AY096422.1 | - | 7 - 8 | 1130 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
