Assay Details
| Gene Symbol: RIC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338629.1 | NP_001319624.1 | 3 - 4 | 361 | 85 | |
| NM_113241.2 | NP_188980.1 | 1 - 2 | 56 | 85 | ||
More Information
Set Membership: |
|

RIC5
Genomic Map