Assay Details
| Gene Symbol: PHO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113246.5 | NP_188985.2 | 5 - 6 | 1192 | 108 | |
| GenBank mRNA | AF474076.1 | - | 5 - 6 | 1021 | 108 | |
More Information
Set Membership: |
|

PHO1
Genomic Map