Assay Details
| Gene Symbol: PHO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113246.5 | NP_188985.2 | 6 - 7 | 1259 | 110 | |
| GenBank mRNA | AF474076.1 | - | 6 - 7 | 1088 | 110 | |
More Information
Set Membership: |
|

PHO1
Genomic Map