Assay Details
| Gene Symbol: GAE6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113287.3 | NP_189024.1 | 1 | 2164 | 82 | |
| GenBank mRNA | AY062625.1 | - | 1 | 1879 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAE6| Gene Symbol: GAE6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113287.3 | NP_189024.1 | 1 | 2164 | 82 | |
| GenBank mRNA | AY062625.1 | - | 1 | 1879 | 82 | |
Set Membership: |
|