Assay Details
Gene Symbol: SYP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113342.4 | NP_189078.2 | 1 - 2 | 438 | 94 | |
GenBank mRNA | BT004300.1 | - | 1 - 2 | 338 | 94 | |
BT008604.1 | - | 1 - 2 | 247 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
