Assay Details
Gene Symbol: UBC37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338687.1 | NP_001325555.1 | 1 - 2 | 539 | 114 | |
NM_001338688.1 | NP_001325554.1 | 2 - 3 | 492 | 114 | ||
NM_001338689.1 | NP_001325556.1 | 2 - 3 | 483 | 114 | ||
NM_113362.2 | NP_566751.1 | 3 - 4 | 398 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001338687.1, NM_001338689.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
