Assay Details
Gene Symbol: RIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338741.1 | NP_001325873.1 | 1 - 2 | 1208 | 149 | |
NM_113411.3 | NP_189143.2 | 2 - 3 | 806 | 149 | ||
GenBank mRNA | AK117488.1 | - | 2 - 3 | 477 | 149 | |
BT024595.1 | - | 1 - 2 | 416 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
