Assay Details
| Gene Symbol: QUA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113418.5 | NP_189150.1 | 1 - 2 | 402 | 122 | |
| GenBank mRNA | AY099574.1 | - | 1 - 2 | 199 | 122 | |
| BT010393.1 | - | 1 - 2 | 162 | 122 | ||
| KJ138811.1 | - | 1 - 2 | 162 | 122 | ||
More Information
Set Membership: |
|

QUA1
Genomic Map