Assay Details
| Gene Symbol: QUA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113418.5 | NP_189150.1 | 2 - 3 | 1642 | 74 | |
| GenBank mRNA | AY099574.1 | - | 2 - 3 | 1439 | 74 | |
| BT010393.1 | - | 2 - 3 | 1402 | 74 | ||
| KJ138811.1 | - | 2 - 3 | 1402 | 74 | ||
More Information
Set Membership: |
|

QUA1
Genomic Map