Assay Details
| Gene Symbol: MAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338764.1 | NP_001326796.1 | 3 - 4 | 394 | 91 | |
| NM_113473.3 | NP_189202.1 | 3 - 4 | 396 | 91 | ||
| GenBank mRNA | BT024875.1 | - | 3 - 4 | 374 | 91 | |
More Information
Set Membership: |
|

MAP1B
Genomic Map