Assay Details
| Gene Symbol: LTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113489.3 | NP_189215.1 | 4 - 5 | 1457 | 84 | |
| GenBank mRNA | AF066079.1 | - | 4 - 5 | 1316 | 84 | |
| AY037262.1 | - | 4 - 5 | 1330 | 84 | ||
| BT002343.1 | - | 4 - 5 | 1258 | 84 | ||
More Information
Set Membership: |
|

LTA2
Genomic Map