Assay Details
| Gene Symbol: AT3G26610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113570.3 | NP_189293.1 | 5 - 6 | 939 | 91 | |
| GenBank mRNA | AK117620.1 | - | 5 - 6 | 937 | 91 | |
More Information
Set Membership: |
|

AT3G26610
Genomic Map