Assay Details
| Gene Symbol: PES2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113596.2 | NP_566801.1 | 12 - 13 | 1819 | 147 | |
| GenBank mRNA | AF360145.1 | - | 12 - 13 | 1816 | 147 | |
| AY056370.1 | - | 12 - 13 | 1802 | 147 | ||
More Information
Set Membership: |
|

PES2
Genomic Map