Assay Details
| Gene Symbol: AT5G63080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125702.4 | NP_201113.2 | 6 - 7 | 936 | 74 | |
| GenBank mRNA | AK176202.1 | - | 6 - 7 | 886 | 74 | |
| BT023460.1 | - | 6 - 7 | 868 | 74 | ||
More Information
Set Membership: |
|

AT5G63080
Genomic Map