Assay Details
| Gene Symbol: HDA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125705.4 | NP_201116.1 | 1 - 2 | 660 | 87 | |
| GenBank mRNA | AY072201.1 | - | 1 - 2 | 615 | 87 | |
| AY142660.1 | - | 1 - 2 | 540 | 87 | ||
More Information
Set Membership: |
|

HDA6
Genomic Map