Assay Details
Gene Symbol: ADK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037052.2 | NP_001032129.1 | 3 - 4 | 480 | 70 | |
NM_125735.4 | NP_201145.1 | 3 - 4 | 480 | 70 | ||
GenBank mRNA | AY039945.1 | - | 3 - 4 | 404 | 70 | |
AY079340.1 | - | 3 - 4 | 390 | 70 | ||
AY085188.1 | - | 3 - 4 | 437 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
