Assay Details
| Gene Symbol: ADK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037052.2 | NP_001032129.1 | 5 - 6 | 706 | 76 | |
| NM_125735.4 | NP_201145.1 | 5 - 6 | 737 | 76 | ||
| GenBank mRNA | AY039945.1 | - | 5 - 6 | 661 | 76 | |
| AY079340.1 | - | 5 - 6 | 647 | 76 | ||
| AY085188.1 | - | 5 - 6 | 694 | 76 | ||
More Information
Set Membership: |
|

ADK1
Genomic Map