Assay Details
| Gene Symbol: UVR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125781.4 | NP_201191.1 | 4 - 5 | 908 | 70 | |
| GenBank mRNA | AY125497.1 | - | 510 | 70 | ||
| BT000600.1 | - | 3 - 4 | 427 | 70 | ||
More Information
Set Membership: |
|

UVR8
Genomic Map