Assay Details
| Gene Symbol: FCLY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125787.5 | NP_201196.1 | 3 - 4 | 1168 | 77 | |
| GenBank mRNA | AY081255.1 | - | 3 - 4 | 1077 | 77 | |
| AY128716.1 | - | 3 - 4 | 1067 | 77 | ||
More Information
Set Membership: |
|

FCLY
Genomic Map