Assay Details
| Gene Symbol: EMB3144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125854.4 | NP_201263.2 | 7 - 8 | 1592 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMB3144| Gene Symbol: EMB3144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125854.4 | NP_201263.2 | 7 - 8 | 1592 | 92 | |
Set Membership: |
|