Assay Details
| Gene Symbol: CDKC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085328.2 | NP_001078797.1 | 2 - 3 | 508 | 109 | |
| NM_125895.5 | NP_201301.1 | 3 - 4 | 410 | 109 | ||
| GenBank mRNA | AF360134.1 | - | 3 - 4 | 395 | 109 | |
| AY039990.1 | - | 3 - 4 | 287 | 109 | ||
More Information
Set Membership: |
|

CDKC2
Genomic Map