Assay Details
| Gene Symbol: MAF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085330.2 | NP_001078799.2 | 4 - 5 | 522 | 122 | |
| NM_001345667.1 | NP_001318879.1 | 3 - 4 | 205 | 122 | ||
More Information
Set Membership: |
|

MAF5
Genomic Map