Assay Details
| Gene Symbol: UBP17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345687.1 | NP_001332735.1 | 5 - 6 | 1431 | 119 | |
| NM_125943.2 | NP_201348.1 | 5 - 6 | 1103 | 119 | ||
More Information
Set Membership: |
|

UBP17
Genomic Map