Assay Details
| Gene Symbol: AT5G65900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125987.3 | NP_201391.1 | 7 - 8 | 1578 | 66 | |
| GenBank mRNA | AY059930.1 | - | 7 - 8 | 1535 | 66 | |
| BT001188.1 | - | 7 - 8 | 1411 | 66 | ||
More Information
Set Membership: |
|

AT5G65900
Genomic Map