Assay Details
| Gene Symbol: MLO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123796.1 | NP_001117268.1 | 12 - 13 | 1711 | 92 | |
| NM_001331977.1 | NP_001320358.1 | 12 - 13 | 1320 | 92 | ||
| NM_101004.4 | NP_172598.1 | 13 - 14 | 1670 | 92 | ||
| GenBank mRNA | AF369563.1 | - | 13 - 14 | 1370 | 92 | |
| AK226449.1 | - | 13 - 14 | 1623 | 92 | ||
| AY086586.1 | - | 13 - 14 | 1581 | 92 | ||
More Information
Set Membership: |
|

MLO2
Genomic Map