Assay Details
| Gene Symbol: MLO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123796.1 | NP_001117268.1 | 2 - 3 | 488 | 100 | |
| NM_001331977.1 | NP_001320358.1 | 1 - 2 | 186 | 100 | ||
| NM_101004.4 | NP_172598.1 | 2 - 3 | 536 | 100 | ||
| GenBank mRNA | AF369563.1 | - | 2 - 3 | 236 | 100 | |
| AK226449.1 | - | 2 - 3 | 489 | 100 | ||
| AY086586.1 | - | 2 - 3 | 447 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001331977.1 transcripts.
Set Membership: |
|

MLO2
Genomic Map